Orders LinkContactUsLinkComponent
Pipette tips for sample preparation, ZipTip®
Pipette tips for sample preparation, ZipTip®
With 0,6 µl C4 resin, standard bed, 10 µl, -, 31 mm, Sample preparation in Sartorius Proline™ multi-channel pipettor, 96 tips/pack
Catalog # MILFZTC04S096
Supplier:   Merck
Pipette tips for sample preparation, ZipTip®
Catalog # MILFZTC04S096
Supplier:  Merck
Pipette tips for sample preparation, ZipTip®
Pipette tips for sample preparation, ZipTip®
With 0,6 µl C4 resin, standard bed, 10 µl, -, 31 mm, Sample preparation in Sartorius Proline™ multi-channel pipettor, 96 tips/pack
Price
423,00 €
Log in for availability
1 * 96 items
Product Details & Documents
ZipTip is a 10 µl pipette tip with a 0,6 or 0,2 µl bed of chromatography media fixed at its end with no dead volume. It is ideal for concentrating and purifying samples for sensitive downstream analyses.
  • Single-step desalting, concentration, and purification
  • Fractionate complex samples for more meaningful data
  • Ideal for peptides, proteins, nucleic acids, and more
  • No dead volume for maximum recovery
  • Eliminates time-consuming chromatography
The ZipTip pipette tip provides a reproducible, high-recovery method for concentrating and purifying femtomoles to picomoles of peptides, proteins and oligonucleotides for improved analytical data.
Available with three different ZipTip resins:
C18 Resin (0,6 µl or 0,2 µl bed volumes): For oligonucleotides or small (<50 kDa) proteins/peptides in aqueous solution
C4 Resin: For proteins or peptides (3 kDa to 100 kDa) in aqueous solution
Strong cationic resin (SCX): For peptides in solution with organic solvents or detergents
Specifications
  • Packed:
    96 tips/pack
  • Sterile:
    -
  • Volume:
    10 µl
  • Length:
    31 mm
  • Tip style:
    With 0,6 µl C4 resin, standard bed
  • For use with:
    Sample preparation in Sartorius Proline™ multi-channel pipettor