Orders LinkContactUsLinkComponent
Salt bridges
Salt bridges
Replacement salt bridge, AL120, double liquid junction, ceramic junction, 138 mm length, for reference electrodes with a ground joint
Catalog # HACHB40A610
Supplier:   Hach
Salt bridges
Catalog # HACHB40A610
Supplier:  Hach
Salt bridges
Salt bridges
Replacement salt bridge, AL120, double liquid junction, ceramic junction, 138 mm length, for reference electrodes with a ground joint
Price
273,00 €
Log in for availability
1 * 1 items
Product Details & Documents
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.
Specifications
  • Description:
    Replacement salt bridge, AL120, double liquid junction, ceramic junction, 138 mm length, for reference electrodes with a ground joint
  • Seq:
    03