Lead Standard Solutions, NIST
Lead, Pb, 10 mg/L, 25 mL
Catalog # HACH2374820
Supplier: Hach
Jump to:
Product Details & Documents
Standard solutions for accuracy checks in lead determinations.
Specifications
Specifications
- Concentration:10 mg/L
- Pk:25 mL
- Element:Lead
- Symbol:Pb