Order Entry
Export
Orders LinkContactUsLinkComponent
Sulphite standard solution

Sulphite standard solution

Sulfite standard solution, 500 ml, 15 mg/l as SO3, 500 mL
Catalog # HACH2408449
Supplier:   Hach
Sulphite standard solution
Catalog # HACH2408449
Supplier:  Hach
CAS Number:  
Sulphite standard solution
Sulphite standard solution

Sulfite standard solution, 500 ml, 15 mg/l as SO3, 500 mL

CAS Number:  
Price
71,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

This equivalent standard is prepared with stabilised sodium thiosulfate, since sulphite is not a stable in solution.

Documents

Specifications

Specifications

  • Concentration:
    15 mg/l as SO<sub>3</sub>
  • Pk:
    500 mL
  • Description:
    Sulfite standard solution, 500 ml

Specifications