Order Entry
Export
Orders LinkContactUsLinkComponent
Aluminium standard solution

Aluminium standard solution

Aluminium, Al, 10 mg/L, 100 mL
Catalog # HACH2305842
Supplier:   Hach
Aluminium standard solution
Catalog # HACH2305842
Supplier:  Hach
CAS Number:  
Aluminium standard solution
Aluminium standard solution

Aluminium, Al, 10 mg/L, 100 mL

CAS Number:  
Price
34,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in aluminium determinations.

Documents

Specifications

Specifications

  • Concentration:
    10 mg/L
  • Pk:
    100 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications