Order Entry
Export
Orders LinkContactUsLinkComponent
Electrode solutions and reagents for ion selective electrodes

Electrode solutions and reagents for ion selective electrodes

Ammonia electrode filling solution, 50 ml, 50 mL
Catalog # HACH4447226
Supplier:   Hach
Electrode solutions and reagents for ion selective electrodes
Catalog # HACH4447226
Supplier:  Hach
CAS Number:  
Electrode solutions and reagents for ion selective electrodes
Electrode solutions and reagents for ion selective electrodes

Ammonia electrode filling solution, 50 ml, 50 mL

CAS Number:  
Price
12,20 €
Log in for availability
1 * 50 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Ammonia electrode filling solution, 50 ml

Specifications