Chlorine standard solution
Chlorine standard solution, 2 ml PourRite ampules (NIST), 25 - 30 mg/l as Cl₂, 20
Supplier: Hach
Jump to:
Product Details & Documents
For accuracy checks in chlorine determinations. Lot specific concentration displayed on label.
- Simple accuracy checks
- Minimise waste
- Helps ensure reliable result
Documents
Specifications
Specifications
- Concentration:25 - 30 mg/l as Cl₂
- Pk:20
- Description:Chlorine standard solution, 2 ml PourRite ampules (NIST)