Order Entry
Export
Orders LinkContactUsLinkComponent
Staining baskets and racks

Staining baskets and racks

Staining basket, for 30 vertical slides, 70×90 mm
Catalog # DELTCP-30
Supplier:   DELTALAB
Staining baskets and racks
Catalog # DELTCP-30
Supplier:  DELTALAB
Staining baskets and racks

Staining basket, for 30 vertical slides, 70×90 mm

Price
106,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Staining baskets and racks, made of stainless steel. Designed for 76×26 mm slides.

  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Description:
    Staining basket, for 30 vertical slides, 70×90 mm
  • Seq:
    02