Orders LinkContactUsLinkComponent
Wire basket, round
Wire basket, round
150×125 mm
Catalog # DELTR-200
Supplier:   DELTALAB
Wire basket, round
Catalog # DELTR-200
Supplier:  DELTALAB
Wire basket, round
Wire basket, round
150×125 mm
Price
102,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
These round baskets feature a folding handle for more convenience. The base of these baskets is reinforced by a thick mesh for added strength.
 : Models with inner crossed dividers are available.
Documents
Specifications
  • Ø×H:
    150×125 mm
  • Seq:
    01