QUANTOFIX® Test Strips, Swimming Pool Test
Swimming pool test 3 in 1, 0 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.2 • 7.6 • 8.4, Free chlorine/ total chlorine: Colour reaction with azines; Alcalinity: Reaction with an acid and mixture of pH indicators; pH: Colour reaction of a pH indicator, 40×40×120 mm (package), 50 test(s), 50
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Semi-qualitative test strips for water qualities in swimming pool.
- Easy dip-and-read procedure Multi-stage colour scale Provides reliable results within 30 seconds No accessories necessary for evaluation Desiccant in the stopper of tube container for optimal protection of the strips against humidity
These strips are used for quick and easy determination of water qualities in swimming pool. The test provides valuable results about the water hardness, the total alkalinity, free and total chlorine and pH.
Specifications
Specifications
- Chemical method:Free chlorine/ total chlorine: Colour reaction with azines; Alcalinity: Reaction with an acid and mixture of pH indicators; pH: Colour reaction of a pH indicator
- Pk:50
- Description:Swimming pool test 3 in 1
- Dimensions:40×40×120 mm (package)
- Tests per kit:50 test(s)
- Range:0 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.2 • 7.6 • 8.4
- Cat. No.:MANA90752
- Parameters:All reagents included
- Contents:50 test strips in a tube
- Platform:Semi-quantitative test strips
- Compliance:Not CE certified and no hazardous material
- Instrument:No
- Shelf life (months):24 (from production)
- Weight:34,1 g



