Sample bottles
30 ml, 30 mm, 33 mm, 76 mm, 24 mm
215-2636
Glass.
- Screw ring
- Different sizes
- Capsules of different formats and materials for a wide variety of applications
Wide neck.
PH30 and PH40 ring.
: Supplied without capsules (to be selected separately).
- Ext.Ø:33 mm
- Thread:30 mm
- Capacity:30 ml
- Height:76 mm
- Neck I-Ø:24 mm



