Order Entry
Export
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Histosette® I, M491

Biopsy processing/embedding cassettes, Histosette® I, M491

Green
Catalog # 720-0900
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Histosette® I, M491
Catalog # 720-0900
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Histosette® I, M491
Biopsy processing/embedding cassettes, Histosette® I, M491

Green

Price
246,00 €
Log in for availability
1 * 1.500 items
Product Details & Documents

Acetal polymer.

  • Resistant to all commonly used histological solvents
  • Square openings (1 mm) to maximise fluid exchange and ensure proper drainage
  • Integral lid
  • Anterior writing area at a 30° angle
Specifications
  • Colour:
    Green
  • Seq:
    03