Order Entry
Export
Orders LinkContactUsLinkComponent
 

Biopsy processing/embedding cassettes, Histosette® I, M491

Pink
  720-0899
Biopsy processing/embedding cassettes, Histosette® I, M491
  720-0899
Biopsy processing/embedding cassettes, Histosette® I, M491
Biopsy processing/embedding cassettes, Histosette® I, M491

Pink

 
246,00 €
   
1 * 1.500 items
 

Acetal polymer.

  • Resistant to all commonly used histological solvents
  • Square openings (1 mm) to maximise fluid exchange and ensure proper drainage
  • Integral lid
  • Anterior writing area at a 30° angle
 
  • Colour:
    Pink
  • Seq:
    02