Orders LinkContactUsLinkComponent
Cylinders, without lid
Cylinders, without lid
100 mm
Catalog # HECH44409024
Cylinders, without lid
Catalog # HECH44409024
Cylinders, without lid
Cylinders, without lid
100 mm
Price
10,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
Specifications
  • Ext.Ø:
    100 mm
  • Seq:
    18