Order Entry
Export
Orders LinkContactUsLinkComponent
 

Aluminium foil sheets

100 mm, 100 mm, 30 µm
  611-9007
 :   Korff
Aluminium foil sheets
  611-9007
 :  Korff
Aluminium foil sheets
Aluminium foil sheets

100 mm, 100 mm, 30 µm

 
82,30 €
   
1 * 1.000 items
 

Aluminium foil, square cut.

  • Economical large packs
  • Tissue paper is inserted between sheets to facilitate handling

Can be used in a wide variety of laboratory tasks.

 
  • Width:
    100 mm
  • Length:
    100 mm
  • Seq:
    02
  • Thickness:
    30 µm