Pipette tips for sample preparation, ZipTip®
With 0,6 µl C18 resin, standard bed, 10 µl, -, 31 mm, Sample preparation in Sartorius Proline™ multi-channel pipettor, 96 tips/pack
Catalog # MILFZTC18S096
Supplier: Merck
Jump to:
Product Details & Documents
ZipTip is a 10 µl pipette tip with a 0,6 or 0,2 µl bed of chromatography media fixed at its end with no dead volume. It is ideal for concentrating and purifying samples for sensitive downstream analyses.
- Single-step desalting, concentration, and purification
- Fractionate complex samples for more meaningful data
- Ideal for peptides, proteins, nucleic acids, and more
- No dead volume for maximum recovery
- Eliminates time-consuming chromatography
The ZipTip pipette tip provides a reproducible, high-recovery method for concentrating and purifying femtomoles to picomoles of peptides, proteins and oligonucleotides for improved analytical data.
Available with three different ZipTip resins:
C18 Resin (0,6 µl or 0,2 µl bed volumes): For oligonucleotides or small (<50 kDa) proteins/peptides in aqueous solution
C4 Resin: For proteins or peptides (3 kDa to 100 kDa) in aqueous solution
Strong cationic resin (SCX): For peptides in solution with organic solvents or detergents
Specifications
- Packed:96 tips/pack
- Sterile:-
- Volume:10 µl
- Length:31 mm
- Tip style:With 0,6 µl C18 resin, standard bed
- For use with:Sample preparation in Sartorius Proline™ multi-channel pipettor



