Hoffmann pinch cocks, heavy type
70 mm
Catalog # BOCH12542
Supplier: BOCHEM
Jump to:
Product Details & Documents
Hoffmann pinch cocks made of nickel plated brass.
- Heavy type
- With flap
Specifications
- Spanning width:70 mm
- Cat. No.:BOCH12542
Hoffmann pinch cocks made of nickel plated brass.