Electron micrography films, Kodak
Product Details & Documents
These micrography films offer considerable latitude in performance and flexibility for most electron microscopy applications. They are suitable for medium-energy electrons (40 to 100 ky) and acceptable results can also be obtained at higher electron levels.
- Engineered to produce high-quality images
- Transmission Electron Micrography (TEM) or electron diffraction studies
- Versatile image capture in the transmission electron microscope for life and material research
Micrography Film 4489 with less curl and shorter pump-down times. Approximately half the speed of Image Film SO-163. Coated on a 0,18 mm Estar support, electron microscope film offers dimensional stability and eliminates the use of glass support.
Image Film SO-163 are twice faster with broad exposure and development ranges. Single film to collect many electrons with improvements in SNR or fewer electrons in exposure-limiting situations (when graininess is a reasonable trade-off for higher speed).
Caution: SO-163 film speed requires longer pump-down times and shows greater film curl. When using SO-163 for the first time, it is recommended to recalibrate the microscope exposure.
Specifications
- Description:Electron microscope film, 4489
- Dimensions:70×70 mm
- Seq:02