Biopsy processing/embedding cassettes, Histosette® II, M486
Grey
720-0243
Acetal polymer. Designed to hold biopsy specimens. Covers are already removed from cassettes and are packed separately in the case.
- Convenient for cassette labelling instruments as the covers are already removed
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
- Colour:Grey