Volumetric flasks, class A, blue markings, BLAUBRAND®
200 ml, 0,15 ± ml
Catalog # 612-2567
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with beaded rim.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
* Wide neck.
Certifications: DIN EN ISO 1042
Ordering information: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
Documents
Technical Note BRAND Volumetric Flasks
Application Notes
Specifications
- Capacity:200 ml
- Seq:06
- Tolerance:0,15 ± ml