Volumetric flasks, class A, blue markings, BLAUBRAND®
100 ml, 0,10 ± ml
612-2566
: Brand
Borosilicate glass 3.3, with beaded rim.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
* Wide neck.
: DIN EN ISO 1042
: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
- Capacity:100 ml
- Seq:05
- Tolerance:0,10 ± ml