Volumetric flasks, class A, blue markings, BLAUBRAND®
50 ml, 0,06 ± ml
612-2565
: Brand
Borosilicate glass 3.3, with beaded rim.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
* Wide neck.
: DIN EN ISO 1042
: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
- Capacity:50 ml
- Seq:04
- Tolerance:0,06 ± ml