Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Histosette® II, M486
Biopsy processing/embedding cassettes, Histosette® II, M486
Green
Catalog # 720-0238
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Histosette® II, M486
Catalog # 720-0238
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Histosette® II, M486
Biopsy processing/embedding cassettes, Histosette® II, M486
Green
Price
195,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Acetal polymer. Designed to hold biopsy specimens. Covers are already removed from cassettes and are packed separately in the case.
  • Convenient for cassette labelling instruments as the covers are already removed
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
Specifications
  • Colour:
    Green