Order Entry
Export
Orders LinkContactUsLinkComponent
Volumetric flasks, class B, Azlon®

Volumetric flasks, class B, Azlon®

50 ml, 14/23, 0,12 ± ml
Catalog # 214-0116
Supplier:   SciLabware
Volumetric flasks, class B, Azlon®
Catalog # 214-0116
Supplier:  SciLabware
Volumetric flasks, class B, Azlon®
Volumetric flasks, class B, Azlon®

50 ml, 14/23, 0,12 ± ml

Price
26,90 €
Log in for availability
1 * 1 items
Product Details & Documents

PP, translucent, with PE stopper.

  • Class B tolerances with a clear easy to read calibration line
  • Feature a mixing chamber at the top of each flask's neck
  • Exposure to temperatures above 60 °C may affect accuracy
  • Cleaning with mild alkaline detergents recommended up to 60 °C

Certifications: ISO 1042

Specifications
  • NS:
    14/23
  • Capacity:
    50 ml
  • Tolerance:
    0,12 ± ml