Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
360 mm, 24/29 NS, 24/29 NS
Catalog # 201-8392
Supplier:   SciLabware
Fractionating columns, Vigreux, Quickfit®
Catalog # 201-8392
Supplier:  SciLabware
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
360 mm, 24/29 NS, 24/29 NS
Price
147,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
No glass jacket. With combination of horizontal and inclined indentations. Horizontal indentations ensure intimate vapour and piquid contatc. Inclined indentations redistribute liquid from walls to centre of column.
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS
  • Usable length:
    360 mm
  • Seq:
    02