Tourniquets
Tourniquet for adults
Catalog # 489-1000
Supplier: TERUMO
Jump to:
Product Details & Documents
These tourniquets are available in three sizes suitable for adults, children or babies.
- Sturdy, safety snap closure with one-hand operation
- Replaceable and washable ribbon
- Metal-, latex- and phthalate-free
Designed for daily use in all clinical settings including ambulance.
Documents
TERUXX-TC_TDS_EN
Datasheet
Specifications
- Description:Tourniquet for adults
- Seq:01