Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cleanibag system, ratiolab®

Cleanibag, White, 6500 ml, 1000 pieces per box, 300×300 mm
  RATI7000260
 :   RATIOLAB
Cleanibag system, ratiolab®
  RATI7000260
 :  RATIOLAB
Cleanibag system, ratiolab®
Cleanibag system, ratiolab®

Cleanibag, White, 6500 ml, 1000 pieces per box, 300×300 mm

 
126,70 €
   
1 * 1.000 items
 

All-purpose PE bags with self-adhesive strip for clinical and laboratory use.

  • Self-adhesive strip: sticks on smooth and clean surfaces (e.g. walls, work benches, beds, bedside tables)
  • Residue-free, removable

 : Other sizes or models available on request, e.g. for intensive care and bags with custom-made imprint.

 : 1000 pieces per cardboard box.

 
  • Packed:
    1000 pieces per box
  • Capacity:
    6500 ml
  • Colour:
    White
  • Description:
    Cleanibag
  • Seq:
    02
  • W×L:
    300×300 mm