Orders LinkContactUsLinkComponent
Disposable base moulds
Disposable base moulds
Base mould, 7×7×5 mm
Catalog # 720-0820
Supplier:   Simport Scientific
Disposable base moulds
Catalog # 720-0820
Supplier:  Simport Scientific
Disposable base moulds
Disposable base moulds
Base mould, 7×7×5 mm
Price
270,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
  • Excellent thermal exchange
  • Smooth interior
  • Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
Delivery information: Each case contains two dispenser boxes of 500 base moulds.
Specifications
  • Description:
    Base mould
  • Dimensions:
    7×7×5 mm
  • Seq:
    01