Orders LinkContactUsLinkComponent
Stoppers, press down, VacuCap®
Red, 16 mm
  720-0555
Stoppers, press down, VacuCap®
  720-0555
Stoppers, press down, VacuCap®
Stoppers, press down, VacuCap®
Red, 16 mm
43,30 €
1 * 1.000 items
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
  • Disposable seals are an inexpensive alternative to standard seals
  • Double flanged seal for an impermeable, secure fit
  • Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.
  • Ø bottom:
    16 mm
  • Colour:
    Red