Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pufferlösung, pH 4.01 Puffer mit Zertifikat, Beutel, 20ml 1 * 25 ST
Catalog # HANNHI70004C_L
Supplier:
Hanna
Expand image
Pufferlösung, pH 4.01 Puffer mit Zertifikat, Beutel, 20ml 1 * 25 ST
Catalog # HANNHI70004C_L
Supplier:
Hanna