Orders LinkContactUsLinkComponent
Conductivity and TDS standards
Conductivity and TDS standards
Conductivity standard 12880 µS/cm (0,1 mol/l; KCl), 25 °C, Sachet, 25
Catalog # HANNHI70030P
Supplier:   Hanna
Conductivity and TDS standards
Catalog # HANNHI70030P
Supplier:  Hanna
CAS Number:  
Conductivity and TDS standards
Conductivity and TDS standards
Conductivity standard 12880 µS/cm (0,1 mol/l; KCl), 25 °C, Sachet, 25
CAS Number:  
Price
53,20 €
Log in for availability
1 * 25 items
Product Details & Documents
An extensive range of standard solutions designed to ensure maximum accuracy of conductivity meters and sensors.
  • The labels on the bottles and sachets include the batch number, expiry date and the relationship between conductivity and temperature
The labels on the bottles and sachets include the batch number, expiry date and the relationship between conductivity and temperature.
Documents
Specifications
Specifications
  • Pack type:
    Sachet
  • Pk:
    25
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 12880 µS/cm (0,1 mol/l; KCl)
Specifications