Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexane ≥99%, GPR RECTAPUR®

Cyclohexane ≥99%, GPR RECTAPUR®

Glass bottle, 2,5 L
Catalog # 23223.324
Flammable
Health
Irritant
Environment
Supplier:   VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Catalog # 23223.324
Supplier:  VWR Chemicals
CAS Number:  110-82-7
Cyclohexane ≥99%, GPR RECTAPUR®
Cyclohexane ≥99%, GPR RECTAPUR®

Glass bottle, 2,5 L

CAS Number:  110-82-7
Price
111,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II
  • REACH:01-2119463273-41
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Substances coloured by H2SO4:
    Passes test
  • Boiling point:
    79 - 81 °C
  • Density (20/4):
    0.778 - 0.779
  • Free acidity:
    Max. 0.0002 meq/g
  • Solidification point:
    5.5 - 6.5 °C
  • Evaporation residue:
    Max. 50 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications