Cyclohexane ≥99%, GPR RECTAPUR®
Glass bottle, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0.7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
- REACH:01-2119463273-41
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Specifications
Specification Test Results
- Assay:Min. 99 %
- IR Spectrum:Passes test
- Substances coloured by H2SO4:Passes test
- Boiling point:79 - 81 °C
- Density (20/4):0.778 - 0.779
- Free acidity:Max. 0.0002 meq/g
- Solidification point:5.5 - 6.5 °C
- Evaporation residue:Max. 50 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L



