Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Chromium (III) oxide analytical reagent 1 * 250 g
Catalog # 22799.235
Supplier:
VWR Chemicals
SDS
Certificates
Expand image
Chromium (III) oxide analytical reagent 1 * 250 g
Catalog # 22799.235
Supplier:
VWR Chemicals
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Packaging Information
750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
SDS
SDS_EN_UK_VWRC22799.pdf
sds_VWRC22799
Product specification
spec_VWRC22799.235
Documentation