ContactUsLinkComponent
Thermometer, mercury, AFNOR STL/0.2/35/85, white backed, measuring range: 35:85/0.2C, length: 375mm, total immersion, durable pigm e nt, with ice point scale, suitable for government verification. Until 10th October 2017 it is still allowed to use 1 * 1 items
Catalog # AMARA411038
Supplier:  Amarell
Thermometer, mercury, AFNOR STL/0.2/35/85, white backed, measuring range: 35:85/0.2C, length: 375mm, total immersion, durable pigm e nt, with ice point scale, suitable for government verification. Until 10th October 2017 it is still allowed to use 1 * 1 items
Catalog # AMARA411038
Supplier:  Amarell