Microplates, 384-well
Black, FLUOTRAC™ 200 medium binding, 384, -
Catalog # 736-0140
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
PS. With the exception of the Small Volume™, all 384-well PS microplates have rounded square wells, which combine the advantages of flexible working volume (10 to 130 µl) with reduced wicking and bubbling. Small Volume™ plates have round wells with a conical geometry, which allows a working volume of just 4 to 25 µl. All plates are supplied without lid, except for some sterile formats.
- Flat bottom well profile
- Choice of solid or µClear® bottom
- Medium or high binding versions available
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Documents
Specifications
- No. of wells:384
- Sterile:-
- Description:Black, FLUOTRAC™ 200 medium binding
- Seq:09
- Cat. No.:736-0140
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact