Immunology tubes
5 ml, round bottom with star, Microlon 200
Catalog # 738-0002
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
PS, clear. Star bottomed tubes have an increased surface area and thus make it possible to bind larger amounts of antigens or antibodies.
Immuno tubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone).
Available with either MICROLON® 200 (medium binding) and MICROLON® 600 (high binding) surfaces.
Ø×L: 12×75 mm
Specifications
- Capacity:5 ml
- Description:round bottom with star, Microlon 200
- Seq:03
- Cat. No.:738-0002
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact



