Order Entry
Export
Orders LinkContactUsLinkComponent
Immunology tubes

Immunology tubes

5 ml, round bottom, Microlon 200
Catalog # 738-0000
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Immunology tubes
Catalog # 738-0000
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Immunology tubes

5 ml, round bottom, Microlon 200

Price
164,00 €
Log in for availability
1 * 2.000 items
Product Details & Documents

PS, clear. Star bottomed tubes have an increased surface area and thus make it possible to bind larger amounts of antigens or antibodies.

Immuno tubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone).

Available with either MICROLON® 200 (medium binding) and MICROLON® 600 (high binding) surfaces.

Ø×L: 12×75 mm

Specifications
  • Capacity:
    5 ml
  • Description:
    round bottom, Microlon 200
  • Seq:
    02
  • Cat. No.:
    738-0000
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact