Order Entry
Export
Orders LinkContactUsLinkComponent
Aluminium foil sheets

Aluminium foil sheets

150 mm, 150 mm, 30 µm
Catalog # 611-9009
Supplier:   Korff
Aluminium foil sheets
Catalog # 611-9009
Supplier:  Korff
Aluminium foil sheets
Aluminium foil sheets

150 mm, 150 mm, 30 µm

Price
122,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents

Aluminium foil, square cut.

  • Economical large packs
  • Tissue paper is inserted between sheets to facilitate handling

Can be used in a wide variety of laboratory tasks.

Specifications
  • Width:
    150 mm
  • Length:
    150 mm
  • Seq:
    04
  • Thickness:
    30 µm