Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonia solution 28%, GPR RECTAPUR®

Ammonia solution 28%, GPR RECTAPUR®

Plastic bottle, fluorinated, 5 L
Catalog # 21182.363
Corrosive
Irritant
Environment
Supplier:   VWR Chemicals
Ammonia solution 28%, GPR RECTAPUR®
Catalog # 21182.363
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution 28%, GPR RECTAPUR®
Ammonia solution 28%, GPR RECTAPUR®

Plastic bottle, fluorinated, 5 L

CAS Number:  1336-21-6
Price
96,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.898 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    28 - 32 %
  • Density (20/4):
    0.880 - 0.910
  • Evaporation residue:
    Max. 0.02 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 5 ppm

Specifications

  • Pack type:
    Plastic bottle, fluorinated
  • Pk:
    5 L

Specifications