Ammonia solution 28%, GPR RECTAPUR®
Glass bottle, 1 L
Supplier: VWR Chemicals
Ammonia solution 28%, GPR RECTAPUR®
Catalog # 21182.294
Supplier: VWR Chemicals
CAS Number: 1336-21-6
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:32…37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.898 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
Specifications
Specification Test Results
- Assay:28 - 32 %
- Density (20/4):0.880 - 0.910
- Evaporation residue:Max. 0.02 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- Fe (Iron):Max. 5 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L



