Order Entry
Export
Orders LinkContactUsLinkComponent
Methanol ≥98.5%, TECHNICAL

Methanol ≥98.5%, TECHNICAL

Plastic drum, 10 L
Catalog # 20903.415
Flammable
Toxic
Health
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Catalog # 20903.415
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  67-56-1
Methanol ≥98.5%, TECHNICAL
Methanol ≥98.5%, TECHNICAL

Plastic drum, 10 L

CAS Number:  67-56-1
Price
99,80 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3 (6.1),II
  • REACH:01-2119433307-44
Specifications

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Evaporation residue:
    Max. 0.015 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L
  • Cat. No.:
    20903.415
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications