Order Entry
Export
Orders LinkContactUsLinkComponent
Stearic acid, powder, TECHNICAL

Stearic acid, powder, TECHNICAL

Plastic bucket, 25 kg
Catalog # 20666.460
Irritant
Supplier:   VWR Chemicals
Stearic acid, powder, TECHNICAL
Catalog # 20666.460
Supplier:  VWR Chemicals
CAS Number:  57-11-4
Stearic acid, powder, TECHNICAL
Stearic acid, powder, TECHNICAL

Plastic bucket, 25 kg

CAS Number:  57-11-4
Price
463,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Boiling Pt:361 °C (1013 hPa)
  • Melting Pt:67 °C
  • Density:0.94 g/cm³ (20 °C)
  • Flash Pt:196 °C (closed cup)
  • Storage Temperature:Refrigerator
  • EINECS:200-313-4
  • REACH:01-2119543894-28
Specifications

Specification Test Results

  • Stearic and palmitic acid:
    Min. 90 %

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications