Stearic acid, powder, TECHNICAL
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Octadecanoic acid, Octadecylic acid
- CAS Number:57-11-4
- Formula:C₁₈H₃₆O₂
- MDL Number:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:13,08882
- Boiling Pt:361 °C (1013 hPa)
- Melting Pt:67 °C
- Density:0.94 g/cm³ (20 °C)
- Flash Pt:196 °C (closed cup)
- Storage Temperature:Refrigerator
- EINECS:200-313-4
- REACH:01-2119543894-28
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Stearic and palmitic acid:Min. 90 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



