Orders LinkContactUsLinkComponent
Multiparameter test kits, model HA-62
Multiparameter test kits, model HA-62
For hardness (as CaCO₃), iron and pH, HA-62B, Hardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 10, 1 KIT
Catalog # HACH183702
Supplier:   Hach
Multiparameter test kits, model HA-62
Catalog # HACH183702
Supplier:  Hach
CAS Number:  
Multiparameter test kits, model HA-62
Multiparameter test kits, model HA-62
For hardness (as CaCO₃), iron and pH, HA-62B, Hardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 10, 1 KIT
CAS Number:  
Price
462,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
  • Popular tests in one rugged kit
  • Uses color disc and drop count titration methods
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    For hardness (as CaCO₃), iron and pH, HA-62B
  • Range:
    Hardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 10
Specifications