Formic acid 98% ≥98%, GPR RECTAPUR®
Plastic bottle, 5 L
: Aminic Acid, Methanoic acid
- :64-18-6
- :HCO₂H
- :MFCD00003297
- :46.03 g/mol
- :100…101 °C (1013 hPa)
- :8,2…8,4 °C
- :1.22 g/cm³ (25 °C)
- :48 °C
- :200-579-1
- :8 (3),II
- :Density: 1.22 g/cm³ (25 °C)
- :Ambient
- :1779
- :01-2119491174-37
- Assay:Min. 98 %
- Refractive index (20°C):1.370 - 1.372
- Acetic acid:Max. 0.2 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 100 ppm
- Evaporation residue:Max. 100 ppm
- Cl (Chloride):Max. 20 ppm
- SO4 (Sulphate):Max. 50 ppm
- Pack type:Plastic bottle
- Pk:5 L



