Order Entry
Export
Orders LinkContactUsLinkComponent
 

Formic acid 98% ≥98%, GPR RECTAPUR®

Glass bottle, Safebreak®, 2,5 L
Catalog # 20320.320
Flammable
Corrosive
Supplier:   VWR Chemicals
Formic acid 98% ≥98%, GPR RECTAPUR®
Catalog # 20320.320
Supplier:  VWR Chemicals
CAS Number:  64-18-6
Formic acid 98% ≥98%, GPR RECTAPUR®
Formic acid 98% ≥98%, GPR RECTAPUR®

Glass bottle, Safebreak®, 2,5 L

CAS Number:  64-18-6
Price
77,80 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Aminic Acid, Methanoic acid
  • CAS Number:64-18-6
  • Formula:HCO₂H
  • MDL Number:MFCD00003297
  • Molecular Weight:46.03 g/mol
  • Boiling Pt:100…101 °C (1013 hPa)
  • Melting Pt:8,2…8,4 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:48 °C
  • EINECS:200-579-1
  • ADR:8 (3),II
  •  :Density: 1.22 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:1779
  • REACH:01-2119491174-37
Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Refractive index (20°C):
    1.370 - 1.372
  • Acetic acid:
    Max. 0.2 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Evaporation residue:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 50 ppm

Specifications

  • Pack type:
    Glass bottle, Safebreak®
  • Pk:
    2,5 L

Specifications