Formic acid 80% ≥80%, TECHNICAL
Glass bottle, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
- UN:3412
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC20315.366_SPEC_EN.pdf P20_VWRC20315.446_SPEC_EN.pdf spec_VWRC20315.297 spec_VWRC20315.297 spec_VWRC20315.366
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:79 - 81 %
- n 20/D:1.369 - 1.372
Specifications
- Pack type:Glass bottle
- Pk:1 L



