Order Entry
Export
Orders LinkContactUsLinkComponent
Acetone ≥99.5%, GPR RECTAPUR®

Acetone ≥99.5%, GPR RECTAPUR®

Plastic bottle, 1 L
Catalog # 20065.293
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Catalog # 20065.293
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone ≥99.5%, GPR RECTAPUR®
Acetone ≥99.5%, GPR RECTAPUR®

Plastic bottle, 1 L

CAS Number:  67-64-1
Price
27,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specification Test Results

  • Related substances:
    Passes test
  • Boiling point:
    56 - 56.7 °C
  • Density (20/4):
    0.79 - 0.793
  • Evaporation residue:
    Max. 20 ppm
  • Assay:
    Min. 99.5 %
  • Acidity:
    Passes test
  • Alkalinity:
    Passes test
  • Appearance of solution:
    Passes test
  • Insolubility in water:
    Passes test
  • IR Spectrum:
    Passes test
  • Substances reducing KMnO4 (as O):
    Max. 2 ppm
  • Water:
    Max. 0.3 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Cat. No.:
    20065.293
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications