Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pH - rolls in paper pH 1,0 - 12,0 2 zones with reading scale of 1 pH units Dosatest 1 * 5 Roll
Catalog # 35300.606
Supplier:
VWR Chemicals
SDS
Certificates
Expand image
pH - rolls in paper pH 1,0 - 12,0 2 zones with reading scale of 1 pH units Dosatest 1 * 5 Roll
Catalog # 35300.606
Supplier:
VWR Chemicals
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
sds_VWRC35300
Documentation